DL-Mevalonolactone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D0587-100MG
TCIAD0587-100MGEA 44 EUR
TCIAD0587-100MG
DL-Mevalonolactone ≥98.0% (by GC)
DL-Mevalonolactone
Formule: C₆H₁₀O₃
Kookpunt: 150°C
Smeltpunt: 28°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00006648
CAS nummer: 674-26-0
EINECS: 211-615-0
Merck Index: 14,06163

Order Now

Specification Test Results

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR