3-(Dimethylamino)benzoic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D0646-25G D0646-500G
TCIAD0646-25GEA 35.2 EUR
TCIAD0646-25G TCIAD0646-500G
3-(Dimethylamino)benzoic acid ≥98.0% (by GC, titration analysis)
3-(Dimethylamino)benzoic acid
Formule: C₉H₁₁NO₂
Kookpunt: 298 °C (1013 hPa)
Smeltpunt: 149…152 °C
Dichtheid: 1,26 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002497
CAS nummer: 99-64-9
EINECS: 202-775-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfb6080d6a59ad9416ff52a1bb5cdd5c1051eab0&vid=22838e72d37528e793c0ba27eb7ebe658ffd1213&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR