N,N-Dimethylaniline ≥99.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D0665-25ML D0665-500ML
TCIAD0665-25MLEA 18.7 EUR
TCIAD0665-25ML TCIAD0665-500ML
N,N-Dimethylaniline ≥99.0% (by GC, titration analysis)
N,N-Dimethylaniline
Formule: C₈H₁₁N
Kookpunt: 194°C
Dichtheid: 0,9559 g/cm³ (20 °C)
Vlampunt: 73°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008304
CAS nummer: 121-69-7
EINECS: 204-493-5
UN: 2253
ADR: 6.1,II
Merck Index: 14,03234
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee6f5d1672df6000ac049d36f3a72950fc4ae98a&vid=28230f904d9e056152b1ac5cf4f0ccf8eed62fa0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR