2,6-Xylidine ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Amino-m-xylene, 2,6-Dimethylaniline

D0669-25ML D0669-500ML D0669-100ML
TCIAD0669-25MLEA 17.6 EUR
TCIAD0669-25ML TCIAD0669-500ML TCIAD0669-100ML
2,6-Xylidine ≥99.0% (by GC)
2,6-Xylidine
Formule: C₈H₁₁N
MW: 121.18 g/mol
Kookpunt: 216°C
Vlampunt: 91°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007747
CAS nummer: 87-62-7
EINECS: 201-758-7
UN: 1711
ADR: 6.1 II
Merck Index: 12,10217
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a585f2e6f2768bce5a564d6c5bd5b78229a6bc17&vid=a2334de99247c5855c9acd23396435a60b085db9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR