Sodium-2,4-dimethylbenzenesulphonate monohydrate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: m-Xylene-4-sulphonic acid Sodium Salt, Sodium m-Xylene-4-sulphonate, 2,4-Dimethylbenzenesulphonic acid Sodium Salt

D0682-25G D0682-500G
TCIAD0682-25GEA 17.6 EUR
TCIAD0682-25G TCIAD0682-500G
Sodium-2,4-dimethylbenzenesulphonate monohydrate ≥98.0% (by titrimetric analysis)
Sodium-2,4-dimethylbenzenesulphonate monohydrate
Formule: C₈H₁₀O₃S
MW: 208.21 g/mol
MDL Nummer: MFCD00041883
CAS nummer: 142063-30-7
EINECS: 212-567-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0c594a3b45055137fa32b377a8af31be254a7cc&vid=438cc84fe9b7e483da78b855ac7ca33bfb432efc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 % (as Monohydrate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR