Benzyl(dimethyl)amine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

D0688-25ML D0688-500ML
TCIAD0688-25MLEA 17.6 EUR
TCIAD0688-25ML TCIAD0688-500ML
Benzyl(dimethyl)amine ≥98.0% (by GC)
Benzyl(dimethyl)amine
Formule: C₆H₅CH₂N(CH₃)₂
Kookpunt: 180°C
Smeltpunt: –75°C
Dichtheid: 0,915 g/cm³ (20 °C)
Vlampunt: 57°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008329
CAS nummer: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07455d8ea132474ef40eba608506f8b1906d4146&vid=07455d8ea132474ef40eba608506f8b1906d4146&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR