N,N'-Dimethylethylendiamine ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D0720-25ML D0720-5ML
TCIAD0720-25MLEA 81.4 EUR
TCIAD0720-25ML TCIAD0720-5ML
N,N'-Dimethylethylendiamine ≥97.0% (by GC, titration analysis)
N,N'-Dimethylethylendiamine
Formule: C₄H₁₂N₂
Kookpunt: 119 °C (1013 hPa)
Dichtheid: 0,82 g/cm³ (20 °C)
Vlampunt: 26°C
MDL Nummer: MFCD00008290
CAS nummer: 110-70-3
EINECS: 203-793-3
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ce2bfb138a409ad0829e55dad61c325d2c43ea1&vid=2ce2bfb138a409ad0829e55dad61c325d2c43ea1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR