Barium diphenylamine-4-sulphonate ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D0876-25G D0876-5G
TCIAD0876-25GEA 103 EUR
TCIAD0876-25G TCIAD0876-5G
Barium diphenylamine-4-sulphonate ≥98.0% (by total nitrogen basis)
Barium diphenylamine-4-sulphonate
Formule: C₂₄H₂₀BaN₂O₆S₂
Smeltpunt: 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007497
CAS nummer: 6211-24-1
EINECS: 228-278-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,07313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ae72a758370870c065973fbd83c1f929499c89a&vid=3ae72a758370870c065973fbd83c1f929499c89a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light gray to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot Water almost transparency
Drying loss max. 1.0 %
Sensitiveness to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR