1,3-Difenylguanidine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D0892-25G D0892-500G
TCIAD0892-25GEA 20.9 EUR
TCIAD0892-25G TCIAD0892-500G
1,3-Difenylguanidine ≥98.0% (by titrimetric analysis)
1,3-Difenylguanidine
Formule: C₁₃H₁₃N₃
Kookpunt: 321 °C (1013 hPa)
Smeltpunt: 146…147 °C
Dichtheid: 1,19 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001758
CAS nummer: 102-06-7
EINECS: 203-002-1
UN: 3077
ADR: 9,III
Merck Index: 14,03324
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8adc7b94b0eea1015d1ceae98fd07e95d2ab42d7&vid=8adc7b94b0eea1015d1ceae98fd07e95d2ab42d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 152.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR