1,3-Diphenylacetone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D0911-25G D0911-500G D0911-100G
TCIAD0911-25GEA 31.9 EUR
TCIAD0911-25G TCIAD0911-500G TCIAD0911-100G
1,3-Diphenylacetone ≥97.0% (by GC)
1,3-Diphenylacetone
Formule: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
Kookpunt: 330 °C (1013 hPa)
Smeltpunt: 30…34 °C
Dichtheid: 1,195 g/cm³ (0 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004795
CAS nummer: 102-04-5
EINECS: 203-000-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdd1a633815c0223978700421f3dfc5f06566cf5&vid=e50d448cc394a36487b427b88a0a9b5057e59988&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR