1,8-Dibromooctane ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D1064-25G D1064-500G D1064-100G
TCIAD1064-25GEA 40.7 EUR
TCIAD1064-25G TCIAD1064-500G TCIAD1064-100G
1,8-Dibromooctane ≥97.0% (by GC)
1,8-Dibromooctane
Formule: BrH₂C(CH₂)₆CH₂Br
Kookpunt: 272°C
Smeltpunt: 15 °C
Dichtheid: 1,46 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000277
CAS nummer: 4549-32-0
EINECS: 224-912-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2babef5f3080e752987711d4e2e9d8469d3343b4&vid=b471269c8f3c4e9fad43d24d7f70bf8edcacbf2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR