Dithiothreitol (DTT, Cleland's reagent) ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D1071-1G D1071-25G D1071-5G
TCIAD1071-1GEA 36.3 EUR
TCIAD1071-1G TCIAD1071-25G TCIAD1071-5G
Dithiothreitol (DTT, Cleland's reagent) ≥98.0% (by titrimetric analysis)
Dithiothreitol (DTT, Cleland's reagent)
Formule: C₄H₁₀O₂S₂
Kookpunt: 130°C
Smeltpunt: 42…44 °C
Vlampunt: ≥ 110 °C ()
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004877
CAS nummer: 3483-12-3
EINECS: 222-468-7
Merck Index: 14,03376
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd1523edc87c93981d015610c18a5add7a94a23&vid=0bd1523edc87c93981d015610c18a5add7a94a23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Melting point 41.0 to 45.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR