2-Decyn-1-ol ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D1264-25ML D1264-5ML
TCIAD1264-25MLEA 651 EUR
TCIAD1264-25ML TCIAD1264-5ML
2-Decyn-1-ol ≥95.0% (by GC)
2-Decyn-1-ol
Formule: C₁₀H₁₈O
Kookpunt: 113°C
MDL Nummer: MFCD00041567
CAS nummer: 4117-14-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c947d7850c4de0e6f3af90ee0cab1c009bacfc3b&vid=c947d7850c4de0e6f3af90ee0cab1c009bacfc3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR