(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (R)-(-)-1,2-O-Isopropylidenglycerol, (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (R)-(-)-2,3-O-Isopropylideneglycerol, (R)-(-)-1,2-O-Isopropylideneglycerol, (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol

D1705-1G D1705-25G D1705-5G
TCIAD1705-1GEA 23.1 EUR
TCIAD1705-1G TCIAD1705-25G TCIAD1705-5G
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97.0% (by GC)
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Formule: C₆H₁₂O₃
MW: 132.16 g/mol
Kookpunt: 78°C
Vlampunt: 80°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003213
CAS nummer: 14347-78-5
EINECS: 405-320-8
Merck Index: 14,05213
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6f10420a1480606f5f84fa545f52448d2e861cc&vid=f6f10420a1480606f5f84fa545f52448d2e861cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Optical purity (GC) min. 98.0 ee%
Specific rotation [a]20/D -11.0 to -15.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR