Ethyl-4-(dimethylamino)benzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 4-(Dimethylamino)benzoic acid ethyl ester, Ethyl 4-dimethylaminobenzoate

D1744-500G D1744-25G D1744-100G
TCIAD1744-500GEA 171 EUR
TCIAD1744-500G TCIAD1744-25G TCIAD1744-100G
Ethyl-4-(dimethylamino)benzoate ≥98.0% (by GC)
Ethyl-4-(dimethylamino)benzoate
Formule: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Kookpunt: 112
MDL Nummer: MFCD00009115
CAS nummer: 10287-53-3
EINECS: 233-634-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ecd96d9ef54f45aae1f046a1bb76e6985e39ce&vid=23ecd96d9ef54f45aae1f046a1bb76e6985e39ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR