2,4-Dichlorobenzyl alcohol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: (2,4-Dichlorophenyl)methanol

D1837-25G
TCIAD1837-25GEA 25.3 EUR
TCIAD1837-25G
2,4-Dichlorobenzyl alcohol ≥97.0% (by GC)
2,4-Dichlorobenzyl alcohol
Formule: C₇H₆Cl₂O
MW: 177.03 g/mol
Kookpunt: 150°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004606
CAS nummer: 1777-82-8
EINECS: 217-210-5
Merck Index: 14,03060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d09a5848329c6ca214edabfd222ae7226b60262d&vid=d09a5848329c6ca214edabfd222ae7226b60262d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 59.0 to 61.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR