2,6-Dichloropyrazine ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D1867-25G D1867-5G
TCIAD1867-25GEA 238 EUR
TCIAD1867-25G TCIAD1867-5G
2,6-Dichloropyrazine ≥99.0% (by GC)
2,6-Dichloropyrazine
Formule: C₄H₂Cl₂N₂
Kookpunt: 122°C
Smeltpunt: 57…58 °C
Dichtheid: 1,713 g/cm³ (20 °C)
MDL Nummer: MFCD00006125
CAS nummer: 4774-14-5
EINECS: 225-316-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8338b23f3d14fb90fc280984122d54fc610acf8b&vid=8338b23f3d14fb90fc280984122d54fc610acf8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 53.0 to 56.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR