2,5-Dimethoxyaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D1893-25G D1893-500G
TCIAD1893-25GEA 30.8 EUR
TCIAD1893-25G TCIAD1893-500G
2,5-Dimethoxyaniline ≥98.0% (by GC)
2,5-Dimethoxyaniline
Formule: C₈H₁₁NO₂
MW: 153.18 g/mol
Kookpunt: 159°C
Smeltpunt: 81
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008368
CAS nummer: 102-56-7
EINECS: 203-040-9
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d76b81a354daa3c0249fc0c011c3e5b8cd349b0&vid=dc77e8e93e4890c586c199b3bedd7dce3610d70f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR