3,5-Dichlorothiophenol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D1905-25G D1905-5G
TCIAD1905-25GEA 312 EUR
TCIAD1905-25G TCIAD1905-5G
3,5-Dichlorothiophenol ≥97.0% (by GC)
3,5-Dichlorothiophenol
Formule: C₆H₄Cl₂S
Kookpunt: 114°C
MDL Nummer: MFCD00041423
CAS nummer: 17231-94-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c5659dcca237bc98e14637a15aaf1b6e94f8f1f&vid=fa040c4c475eacec1aa736bb252c4c67e03a32a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 62.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR