3',4'-Difluoroacetophenone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-(3,4-Difluorophenyl)ethan-1-one, 1-(3,4-Difluorophenyl)ethanone

D1906-25G D1906-5G
TCIAD1906-25GEA 113 EUR
TCIAD1906-25G TCIAD1906-5G
3',4'-Difluoroacetophenone ≥97.0% (by GC)
3',4'-Difluoroacetophenone
Formule: C₈H₆F₂O
MW: 156.13 g/mol
Kookpunt: 94°C
Smeltpunt: 20
Dichtheid: 1.24
Vlampunt: 75°C
MDL Nummer: MFCD00009891
CAS nummer: 369-33-5
EINECS: 206-717-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e217324d0aee2c7efcb8368f23438ada112f8551&vid=e217324d0aee2c7efcb8368f23438ada112f8551&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR