1,3-Diiminoisoindoline ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D2079-25G D2079-5G
TCIAD2079-25GEA 102 EUR
TCIAD2079-25G TCIAD2079-5G
1,3-Diiminoisoindoline ≥98.0% (by titrimetric analysis)
1,3-Diiminoisoindoline
Formule: C₈H₇N₃
MDL Nummer: MFCD00010062
CAS nummer: 57500-34-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR