trans-2,3-Dibromo-2-butene-1,4-diol ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D2169-25G D2169-500G
TCIAD2169-25GEA 22 EUR
TCIAD2169-25G TCIAD2169-500G
trans-2,3-Dibromo-2-butene-1,4-diol ≥99.0% (by GC)
trans-2,3-Dibromo-2-butene-1,4-diol
Formule: C₄H₆Br₂O₂
MDL Nummer: MFCD00004698
CAS nummer: 21285-46-1
EINECS: 244-313-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20ed3f295eea494beb092ecc6c0c1c489883060c&vid=20ed3f295eea494beb092ecc6c0c1c489883060c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystaline
Purity (GC) min. 99.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR