N,N-Dipropyl-L-alanine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D2224-1G
TCIAD2224-1GEA 90.2 EUR
TCIAD2224-1G
N,N-Dipropyl-L-alanine ≥98.0% (by GC, titration analysis)
N,N-Dipropyl-L-alanine
Formule: C₉H₁₉NO₂
MDL Nummer: MFCD00010823
CAS nummer: 81854-56-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 98.0 deg-C
Specific rotation [a]20/D +14.0 to +18.0 deg (C=2, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR