3,5-Di-tert-butylpyrocatechol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D2432-25G
TCIAD2432-25GEA 97.9 EUR
TCIAD2432-25G
3,5-Di-tert-butylpyrocatechol ≥98.0% (by GC)
3,5-Di-tert-butylpyrocatechol
Formule: C₁₄H₂₂O₂
Kookpunt: 145°C
Smeltpunt: 96…97 °C
MDL Nummer: MFCD00008819
CAS nummer: 1020-31-1
EINECS: 213-816-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 102.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR