1,1-Diphenyl-2-propyn-1-ol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1,1-Diphenylprop-2-yn-1-ol

D2495-25G D2495-5G
TCIAD2495-25GEA 214 EUR
TCIAD2495-25G TCIAD2495-5G
1,1-Diphenyl-2-propyn-1-ol ≥97.0% (by GC)
1,1-Diphenyl-2-propyn-1-ol
Formule: C₁₅H₁₂O
MW: 208.26 g/mol
Kookpunt: 200°C
MDL Nummer: MFCD00041570
CAS nummer: 3923-52-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 47.0 to 51.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR