(-)-Dipivaloyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D2686-25G D2686-5G
TCIAD2686-25GEA 219 EUR
TCIAD2686-25G TCIAD2686-5G
(-)-Dipivaloyl-L-tartaric acid ≥98.0% (by HPLC, titration analysis)
(-)-Dipivaloyl-L-tartaric acid
Formule: C₁₄H₂₂O₈
MDL Nummer: MFCD00066535
CAS nummer: 65259-81-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 147.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR