1,3-Diacetoxyacetone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D2843-25G D2843-5G
TCIAD2843-25GEA 266 EUR
TCIAD2843-25G TCIAD2843-5G
1,3-Diacetoxyacetone ≥98.0% (by GC)
1,3-Diacetoxyacetone
Formule: C₇H₁₀O₅
MDL Nummer: MFCD01310655
CAS nummer: 6946-10-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C
Solubility in Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR