2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D2853-1G D2853-5G
TCIAD2853-1GEA 79.2 EUR
TCIAD2853-1G TCIAD2853-5G
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥98.0% (by GC)
2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formule: C₁₄H₂₁BO₃
MDL Nummer: MFCD02093724
CAS nummer: 269410-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67243d2066238d6cf895bee085ef5efec065c054&vid=67243d2066238d6cf895bee085ef5efec065c054&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR