Dodecaethylene glycol monomethyl ether ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D2904-100MG D2904-1G
TCIAD2904-100MGEA 99 EUR
TCIAD2904-100MG TCIAD2904-1G
Dodecaethylene glycol monomethyl ether ≥90.0% (by GC)
Dodecaethylene glycol monomethyl ether
Formule: C₂₅H₅₂O₁₃
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD06797088
CAS nummer: 5702-16-9

Order Now

Specification Test Results

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR