Diethyl (bromodifluoromethyl)phosphonate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D3071-5G D3071-25G
TCIAD3071-5GEA 38.5 EUR
TCIAD3071-5G TCIAD3071-25G
Diethyl (bromodifluoromethyl)phosphonate ≥97.0% (by GC)
Diethyl (bromodifluoromethyl)phosphonate
Formule: C₅H₁₀BrF₂O₃P
Kookpunt: 102°C
MDL Nummer: MFCD00069118
CAS nummer: 65094-22-6

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR