Diethyl (4-isopropylbenzyl)phosphonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diethyl 4-isopropylbenzylphosphonate

D3325-5G
TCIAD3325-5GEA 338 EUR
TCIAD3325-5G
Diethyl (4-isopropylbenzyl)phosphonate ≥98.0% (by GC)
Diethyl (4-isopropylbenzyl)phosphonate
Formule: C₁₄H₂₃O₃P
MW: 270.300 g/mol
MDL Nummer: MFCD08276362
CAS nummer: 77237-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=480b097d702de0664f128e86b17a3b6f08a15355&vid=480b097d702de0664f128e86b17a3b6f08a15355&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR