3,4-Difluorophenylboronic acid (contains varying a mounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D3350-25G D3350-5G
TCIAD3350-25GEA 102 EUR
TCIAD3350-25G TCIAD3350-5G
3,4-Difluorophenylboronic acid (contains varying a mounts of Anhydride)
3,4-Difluorophenylboronic acid
Formule: C₆H₅BF₂O₂
Smeltpunt: 310°C
MDL Nummer: MFCD00807405
CAS nummer: 168267-41-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR