Dimethyl-L-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D3353-25G D3353-5G
TCIAD3353-25GEA 130 EUR
TCIAD3353-25G TCIAD3353-5G
Dimethyl-L-glutamate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Dimethyl-L-glutamate hydrochloride
Formule: C₇H₁₃NO₄
MDL Nummer: MFCD00038879
CAS nummer: 23150-65-4
EINECS: 245-461-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 91.0 to 101.0 deg-C
Specific rotation [a]20/D +24.0 to +28.0 deg (C=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR