2,4-Dimethyldiphenylamine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: N-Phenyl-2,4-xylidine, 2,4-Dimethyl-N-phenylaniline, Phenyl(2,4-xylyl)amine

D3398-25G
TCIAD3398-25GEA 83.6 EUR
TCIAD3398-25G
2,4-Dimethyldiphenylamine ≥98.0% (by GC)
2,4-Dimethyldiphenylamine
Formule: C₁₄H₁₅N
MW: 197.28 g/mol
Kookpunt: 170°C
MDL Nummer: MFCD08275461
CAS nummer: 25078-04-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystaline
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR