3,5-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D3407-25G D3407-5G
TCIAD3407-25GEA 125 EUR
TCIAD3407-25G TCIAD3407-5G
3,5-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
3,5-Difluorobenzoyl chloride
Formule: C₇H₃ClF₂O
Dichtheid: 1,5 g/cm³ (20 °C)
Vlampunt: 69°C
MDL Nummer: MFCD00010309
CAS nummer: 129714-97-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82063875b0f67cadec2b4f8ac5a06206ad88fccd&vid=82063875b0f67cadec2b4f8ac5a06206ad88fccd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR