Dimethyl trisulphide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D3418-25G D3418-5G
TCIAD3418-25GEA 148 EUR
TCIAD3418-25G TCIAD3418-5G
Dimethyl trisulphide ≥98.0% (by GC)
Dimethyl trisulphide
Formule: C₂H₆S₃
Kookpunt: 170°C
Smeltpunt: -68°C
Vlampunt: 56°C
MDL Nummer: MFCD00039808
CAS nummer: 3658-80-8
EINECS: 222-910-9
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7f54bf56b0d9049f17d986a80eb54fc75f53e81&vid=f7f54bf56b0d9049f17d986a80eb54fc75f53e81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR