Difurfuryl sulphide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D3420-25G D3420-5G
TCIAD3420-25GEA 123 EUR
TCIAD3420-25G TCIAD3420-5G
Difurfuryl sulphide ≥98.0% (by GC)
Difurfuryl sulphide
Formule: C₁₀H₁₀O₂S
Kookpunt: 143°C
MDL Nummer: MFCD00010081
CAS nummer: 13678-67-6
EINECS: 237-172-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5c5b0afbeaf808b174ce5d545939a590fd6f5cc&vid=c5c5b0afbeaf808b174ce5d545939a590fd6f5cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR