4-(Dimethylamino)benzyl alcohol ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D3482-25G D3482-5G
TCIAD3482-25GEA 703 EUR
TCIAD3482-25G TCIAD3482-5G
4-(Dimethylamino)benzyl alcohol ≥98.0% (by titrimetric analysis)
4-(Dimethylamino)benzyl alcohol
Formule: C₉H₁₃NO
Kookpunt: 123°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD03840449
CAS nummer: 1703-46-4

Order Now

Specification Test Results

Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR