(+)-Di-p-anisoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D3490-25G D3490-5G
TCIAD3490-25GEA 91.3 EUR
TCIAD3490-25G TCIAD3490-5G
(+)-Di-p-anisoyl-D-tartaric acid ≥98.0% (by HPLC, titration analysis)
(+)-Di-p-anisoyl-D-tartaric acid
Formule: C₂₀H₁₈O₁₀
MDL Nummer: MFCD07368366
CAS nummer: 191605-10-4
EINECS: 471-150-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR