N,N'-Di-p-tolylbenzidine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D3567-1G D3567-5G
TCIAD3567-1GEA 49.5 EUR
TCIAD3567-1G TCIAD3567-5G
N,N'-Di-p-tolylbenzidine ≥98.0% (by GC)
N,N'-Di-p-tolylbenzidine
Formule: C₂₆H₂₄N₂
MDL Nummer: MFCD09833311
CAS nummer: 10311-61-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 237.0 to 243.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR