2'-Deoxyinosine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D3584-1G D3584-5G
TCIAD3584-1GEA 37.4 EUR
TCIAD3584-1G TCIAD3584-5G
2'-Deoxyinosine ≥98.0% (by HPLC, titration analysis)
2'-Deoxyinosine
Formule: C₁₀H₁₂N₄O₄
Smeltpunt: 220°C
MDL Nummer: MFCD00005762
CAS nummer: 890-38-0
EINECS: 212-964-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa8a87bb81b09ff31f7f511ab494c11f02a59ed7&vid=fa8a87bb81b09ff31f7f511ab494c11f02a59ed7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -29.0 to -33.0 deg (C=1, 0.1mol/L NaOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR