N,N-Dimethylglycine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

D3585-25G D3585-5G
TCIAD3585-25GEA 136 EUR
TCIAD3585-25G TCIAD3585-5G
N,N-Dimethylglycine ≥98.0% (by titrimetric analysis)
N,N-Dimethylglycine
Formule: C₄H₉NO₂
MW: 103.12 g/mol
MDL Nummer: MFCD00004283
CAS nummer: 1118-68-9
EINECS: 214-267-8
Merck Index: 14,03244
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=024a2949a1090a30d2991e2396344e08e3b4304e&vid=024a2949a1090a30d2991e2396344e08e3b4304e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR