Diethyl (4-iodobenzyl)phosphonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D3689-25G D3689-5G
TCIAD3689-25GEA 306 EUR
TCIAD3689-25G TCIAD3689-5G
Diethyl (4-iodobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (4-iodobenzyl)phosphonate
Formule: C₁₁H₁₆IO₃P
MDL Nummer: MFCD09033365
CAS nummer: 173443-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd865cb8feb4786a324ab31a87b19d1fb4f3d824&vid=bd865cb8feb4786a324ab31a87b19d1fb4f3d824&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR