Naphthalene-2,7-dicarbonitrile ≥94.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D3705-1G
TCIAD3705-1GEA 138 EUR
TCIAD3705-1G
Naphthalene-2,7-dicarbonitrile ≥94.0% (by GC)
Naphthalene-2,7-dicarbonitrile
Formule: C₁₂H₆N₂
Smeltpunt: 267°C
CAS nummer: 39718-11-1
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecd4069ee7760c9443132f98ae013f2d25e6717a&vid=ecd4069ee7760c9443132f98ae013f2d25e6717a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 94.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR