1,3-Dicyclohexylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D3850-1G D3850-5G D3850-25G
TCIAD3850-1GEA 58.3 EUR
TCIAD3850-1G TCIAD3850-5G TCIAD3850-25G
1,3-Dicyclohexylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1,3-Dicyclohexylimidazolium tetrafluoroborate
Formule: C₁₅H₂₅BF₄N₂
MDL Nummer: MFCD04973310
CAS nummer: 286014-38-8
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR