EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

D4029-25G D4029-5G D4029-100G
TCIAD4029-25GEA 159 EUR
TCIAD4029-25G TCIAD4029-5G TCIAD4029-100G
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide) ≥98.0% (by GC, titration analysis)
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)
Formule: C₈H₁₇N₃
Kookpunt: 45°C
Smeltpunt: 114…115 °C
Dichtheid: 0,885 g/cm³ (20 °C)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00044916
CAS nummer: 1892-57-5
EINECS: 217-579-2
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd48474d2524b1f91c42b222ffc402c806849c01&vid=cd48474d2524b1f91c42b222ffc402c806849c01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR