2-Amino-3,4-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D4120-5G D4120-25G
TCIAD4120-5GEA 20.9 EUR
TCIAD4120-5G TCIAD4120-25G
2-Amino-3,4-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-3,4-dimethylbenzoic acid
Formule: C₉H₁₁NO₂
MDL Nummer: MFCD00130041
CAS nummer: 50419-58-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR