Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate ≥95.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: [[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonic Acid Diethyl Ester, 9-[2-(Diethoxyphosphorylmethoxy)ethyl]adenine

D4256-1G D4256-5G
TCIAD4256-1GEA 61.6 EUR
TCIAD4256-1G TCIAD4256-5G
Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate ≥95.0% (by HPLC, titration analysis)
Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate
Formule: C₁₂H₂₀N₅O₄P
MW: 329.30 g/mol
MDL Nummer: MFCD07783825
CAS nummer: 116384-53-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR