(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D4418-5G D4418-1G
TCIAD4418-5GEA 451 EUR
TCIAD4418-5G TCIAD4418-1G
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥98.0% (by GC)
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol
Formule: C₂₀H₂₀Br₂O₂
Smeltpunt: 149°C
MDL Nummer: MFCD03093977
CAS nummer: 65355-08-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR