Diethylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

D4643-1G D4643-5G
TCIAD4643-1GEA 44 EUR
TCIAD4643-1G TCIAD4643-5G
Diethylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
Diethylamine hydroiodide
Formule: C₄H₁₁N
CAS nummer: 19833-78-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 173.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR